Documents

52042

Sigma-Aldrich

Hexaethyldisiloxane

≥99.0% (GC)

Linear Formula:
(C2H5)3SiOSi(C2H5)3
CAS Number:
Molecular Weight:
246.54
Beilstein/REAXYS Number:
1750865
EC Number:
MDL number:
PubChem Substance ID:

assay

≥99.0% (GC)

refractive index

n20/D 1.433 (lit.)

bp

233-236 °C (lit.)

density

0.843 g/mL at 25 °C (lit.)

SMILES string

CC[Si](CC)(CC)O[Si](CC)(CC)CC

Looking for similar products? Visit Product Comparison Guide

Other Notes

Poor silylating agent for acids

Certificate of Analysis

Certificate of Origin

H. Matsumoto et al.
Chemistry Letters (Jpn), 1475-1475 (1980)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service