522481

Sigma-Aldrich

Cadmium(II) formate

99.9% trace metals basis

Linear Formula:
(HCO2)2Cd
CAS Number:
Molecular Weight:
202.45
EC Number:
MDL number:
PubChem Substance ID:

assay

99.9% trace metals basis

form

solid

mp

>350 °C (lit.)

SMILES string

[Cd++].[H]C([O-])=O.[H]C([O-])=O

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service