525669

Sigma-Aldrich

2-Nitro-4′-chlorodiphenylamine

98%

Linear Formula:
ClC6H4NHC6H4NO2
CAS Number:
Molecular Weight:
248.67
EC Number:
MDL number:
PubChem Substance ID:

assay

98%

mp

145-148 °C (lit.)

SMILES string

[O-][N+](=O)c1ccccc1Nc2ccc(Cl)cc2

InChI

1S/C12H9ClN2O2/c13-9-5-7-10(8-6-9)14-11-3-1-2-4-12(11)15(16)17/h1-8,14H

InChI key

RCLKXSIRDRWUGX-UHFFFAOYSA-N

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service