Documents

525774

Sigma-Aldrich

Isooctyl thioglycolate

Synonym(s):
IOTG™
Linear Formula:
HSCH2CO2(CH2)5CH(CH3)2
CAS Number:
Molecular Weight:
204.33
EC Number:
MDL number:

SMILES string

CC(C)CCCCCOC(=O)CS

Legal Information

Product of Arkema

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service