526177

Sigma-Aldrich

DBE-2

Synonym(s):
Mixture of dimethyl glutarate and dimethyl adipate, Dibasic ester mixture
Linear Formula:
CH3O2C(CH2)nCO2CH3 (n=3,4)
MDL number:
PubChem Substance ID:

vapor pressure

0.2 mmHg ( 20 °C)

autoignition temp.

698 °F

expl. lim.

8 %

refractive index

n20/D 1.425 (lit.)

bp

210-235 °C (lit.)

mp

−13 °C (lit.)

density

1.081 g/mL at 25 °C (lit.)

SMILES string

COC(=O)CCCC(=O)OC.COC(=O)CCCCC(=O)OC

InChI

1S/C8H14O4.C7H12O4/c1-11-7(9)5-3-4-6-8(10)12-2;1-10-6(8)4-3-5-7(9)11-2/h3-6H2,1-2H3;3-5H2,1-2H3

InChI key

FSCIDASGDAWVED-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

storage_class_code

10 - Combustible liquids

WGK Germany

WGK 1

Flash Point(F)

219.2 °F - closed cup

Flash Point(C)

104 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service