All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C13H10O
CAS Number:
Molecular Weight:
182.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
assay
95%
mp
102-106 °C (lit.)
SMILES string
[H]C(=O)c1ccc2CCc3cccc1c23
InChI
1S/C13H10O/c14-8-11-7-6-10-5-4-9-2-1-3-12(11)13(9)10/h1-3,6-8H,4-5H2
Inchi Key
VNEOTRCBGZQYJT-UHFFFAOYSA-N
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service