527092

Sigma-Aldrich

Diacetoxycumolone

96%

Synonym(s):
4-[2,5-Bis(acetyloxy)-3,4,6-trimethylphenyl]-2-butanone
Empirical Formula (Hill Notation):
C17H22O5
CAS Number:
Molecular Weight:
306.35
MDL number:
PubChem Substance ID:

assay

96%

mp

86-90 °C (lit.)

SMILES string

CC(=O)CCc1c(C)c(OC(C)=O)c(C)c(C)c1OC(C)=O

InChI

1S/C17H22O5/c1-9(18)7-8-15-12(4)16(21-13(5)19)10(2)11(3)17(15)22-14(6)20/h7-8H2,1-6H3

InChI key

ZLPIWEHMRPAXFS-UHFFFAOYSA-N

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service