Documents

52969

Sigma-Aldrich

Boc-α-benzyl-DL-Pro-OH

≥96.0%

Synonym(s):
1-Boc-2-benzyl-2-pyrrolidinecarboxylic acid, Boc-α-benzyl-DL-proline
Empirical Formula (Hill Notation):
C17H23NO4
CAS Number:
Molecular Weight:
305.37
MDL number:
PubChem Substance ID:

assay

≥96.0%

SMILES string

CC(C)(C)OC(=O)N1CCCC1(Cc2ccccc2)C(O)=O

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service