Documents

529842

Sigma-Aldrich

Acetylcyclopentadienylmanganese(I) tricarbonyl

Empirical Formula (Hill Notation):
C10H7MnO4
CAS Number:
Molecular Weight:
246.10
MDL number:
PubChem Substance ID:

mp

41-46 °C (lit.)

SMILES string

[Mn].[C-]#[O+].[C-]#[O+].[C-]#[O+].CC(=O)[C]1[CH][CH][CH][CH]1

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service