Skip to Content
MilliporeSigma

535346

Pentachlorothiophenol

96%

Synonym(s):

Pentachlorobenzenethiol

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Linear Formula:
C6Cl5SH
CAS Number:
Molecular Weight:
282.40
EC Number:
205-107-8
UNSPSC Code:
12352100
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Pentachlorothiophenol, 96%

InChI key

LLMLGZUZTFMXSA-UHFFFAOYSA-N

InChI

1S/C6HCl5S/c7-1-2(8)4(10)6(12)5(11)3(1)9/h12H

SMILES string

Sc1c(Cl)c(Cl)c(Cl)c(Cl)c1Cl

assay

96%

mp

223-227 °C (lit.)

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J To-Figueras et al.
Toxicology letters, 56(1-2), 87-94 (1991-04-01)
The appearance of sex-related differences in the excretion of sulphur derivatives of hexachlorobenzene (HCB) and pentachloronitrobenzene (PCNB) was studied in vitro and in vivo. Sexually immature rats given HCB showed initially no differences in the excretion of N-acetyl-S-(pentachlorophenyl)cysteine, but 5-8
M D'Amour et al.
Toxicology and applied pharmacology, 112(2), 229-234 (1992-02-01)
Hexachlorobenzene (HCB) induces hepatic porphyria and liver cancer in female rats, whereas toxicity is minimal in male rats. HCB is biotransformed to sulfur-containing metabolites originating from conjugation to glutathione (GSH). This study aimed to assess differences in GSH conjugation of
Conversion products of hexachlorobenzene and their role in the disturbance of the porphyrin pathway in rats.
G Koss et al.
The International journal of biochemistry, 12(5-6), 1003-1006 (1980-01-01)
N Legault et al.
Biochemical pharmacology, 54(1), 19-25 (1997-07-01)
Hexachlorobenzene (HCB) is porphyrinogenic in adult female but not in male rats. This study aimed to assess the role of 17beta-estradiol in the induction of porphyria by HCB in both sexes by adding or removing the hormone. Groups of intact
Ramadan A Abuknesha et al.
Analytical and bioanalytical chemistry, 379(3), 411-418 (2004-04-23)
A polyclonal antiserum to pentachlorothiophenol-acetic acid-KLH was generated in sheep and assessed by solid phase ELISA. The assessment procedure included use of double checkerboard analysis in the absence and in the presence of analyte loads, estimation of cross reactivities of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service