Skip to Content
MilliporeSigma

540579

Sigma-Aldrich

(S)-(−)-5,5′,6,6′,7,7′,8,8′-Octahydro-1,1′−bi-2-naphthol

optical purity ee: 99% (HPLC), 97%

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

About This Item

Empirical Formula (Hill Notation):
C20H22O2
CAS Number:
Molecular Weight:
294.39
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.21

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

assay

97%

optical activity

[α]20/D −75°, c = 1 in THF

optical purity

ee: 99% (HPLC)

mp

165-169 °C (lit.)

SMILES string

Oc1c(c4c(cc1)CCCC4)c2c3c(ccc2O)CCCC3

InChI

1S/C20H22O2/c21-17-11-9-13-5-1-3-7-15(13)19(17)20-16-8-4-2-6-14(16)10-12-18(20)22/h9-12,21-22H,1-8H2

InChI key

UTXIFKBYNJRJPH-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service