MilliporeSigma
All Photos(1)

541532

Sigma-Aldrich

4-Fluoro-1,3-diaminobenzene

95%

Synonym(s):
4-Fluoro-1,3-phenylenediamine
Linear Formula:
FC6H3(NH2)2
CAS Number:
Molecular Weight:
126.13
EC Number:
MDL number:
PubChem Substance ID:

Quality Level

assay

95%

bp

152-153 °C (lit.)

mp

34-37 °C (lit.)

SMILES string

Nc1ccc(F)c(N)c1

InChI

1S/C6H7FN2/c7-5-2-1-4(8)3-6(5)9/h1-3H,8-9H2

InChI key

QNDFYLBDUWCFJO-UHFFFAOYSA-N

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service