544116

Sigma-Aldrich

2,2,4,4-Tetrafluoro-1,3-dithietane

97%

Empirical Formula (Hill Notation):
C2F4S2
CAS Number:
Molecular Weight:
164.15
EC Number:
MDL number:
PubChem Substance ID:

assay

97%

refractive index

n20/D 1.393 (lit.)

bp

49-50 °C (lit.)

density

1.582 g/mL at 25 °C (lit.)

SMILES string

FC1(F)SC(F)(F)S1

InChI

1S/C2F4S2/c3-1(4)7-2(5,6)8-1

InChI key

MPOCEMBRPHOCMK-UHFFFAOYSA-N

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service