Skip to Content
MilliporeSigma

544191

Nickel(II) stearate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
Ni(H3C(CH2)16CO2)2
CAS Number:
Molecular Weight:
625.63
UNSPSC Code:
12352300
PubChem Substance ID:
EC Number:
218-744-1
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


mp

82-86 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCC(=O)O[Ni]OC(=O)CCCCCCCCCCCCCCCCC



pictograms

Health hazardEnvironment

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A Inhalation - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

325.0 °F - closed cup

flash_point_c

162.8 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library