Documents

544345

Sigma-Aldrich

(Dimethylvinylsilyloxy)heptacyclopentyltricycloheptasiloxanediol

Empirical Formula (Hill Notation):
C39H74O12Si8
CAS Number:
Molecular Weight:
959.68
MDL number:
PubChem Substance ID:

mp

164-167 °C (lit.)

SMILES string

C[Si](C)(O[Si]12O[Si](O[Si]3(O)O[Si](O)(O[Si](O3)(C4CCCC4)C5CCCC5)O[Si](O1)(C6CCCC6)C7CCCC7)(O[Si](O2)(C8CCCC8)C9CCCC9)C%10CCCC%10)C=C

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service