547204

Sigma-Aldrich

1,1-Dimethoxyoctane

97%

Linear Formula:
H3C(CH2)6CH(OCH3)2
CAS Number:
Molecular Weight:
174.28
EC Number:
MDL number:
PubChem Substance ID:
Pricing and availability is not currently available.

assay

97%

refractive index

n20/D 1.416 (lit.)

bp

206-207 °C (lit.)

density

0.851 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCC(OC)OC

Looking for similar products? Visit Product Comparison Guide

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

WGK Germany

2

Flash Point(F)

194 °F

Flash Point(C)

90 °C

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.