551155

Sigma-Aldrich

(R)-(+)-2-Amino-3-methyl-1,1-diphenylbutane

98%

Empirical Formula (Hill Notation):
C17H21N
CAS Number:
Molecular Weight:
239.36
MDL number:
PubChem Substance ID:

assay

98%

optical activity

[α]20/D +6.5°, c = 1 in chloroform

mp

73-76 °C (lit.)

SMILES string

CC(C)[C@@H](N)C(c1ccccc1)c2ccccc2

Legal Information

A Product of Onyx Scientific, U.K.

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service