552623

Sigma-Aldrich

3-(4-Nitrobenzamido)propyl-functionalized silica gel

200-400 mesh, extent of labeling: 1.2 mmol/g loading

MDL number:
PubChem Substance ID:

extent of labeling

1.2 mmol/g loading

particle size

200-400 mesh

pore size

60 Å pore size

surface area

500 m2/g

SMILES string

O=[SiH2].CCCNC(=O)c1ccc(cc1)[N+]([O-])=O

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service