Documents

552941

Sigma-Aldrich

Calcium carbonate

random crystals, optical grade

Linear Formula:
CaCO3
CAS Number:
Molecular Weight:
100.09
Beilstein/REAXYS Number:
8008338
EC Number:
MDL number:

grade

optical grade

form

random crystals

density

2.93 g/mL at 25 °C (lit.)

SMILES string

[Ca++].[O-]C([O-])=O

InChI

1S/CH2O3.Ca/c2-1(3)4;/h(H2,2,3,4);/q;+2/p-2

InChI key

VTYYLEPIZMXCLO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service