Skip to Content
MilliporeSigma

557587

5,12-Dihydro-5,12-dimethylquino[2,3-b]acridine-7,14-dione

Synonym(s):

DMQA

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C22H16N2O2
CAS Number:
Molecular Weight:
340.37
NACRES:
NA.23
PubChem Substance ID:
UNSPSC Code:
12352103
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

InChI

1S/C22H16N2O2/c1-23-17-9-5-3-7-13(17)21(25)15-12-20-16(11-19(15)23)22(26)14-8-4-6-10-18(14)24(20)2/h3-12H,1-2H3

SMILES string

CN1c2ccccc2C(=O)c3cc4N(C)c5ccccc5C(=O)c4cc13

InChI key

SCZWJXTUYYSKGF-UHFFFAOYSA-N

mp

286 °C (dec.) (lit.)

λmax

295 nm in methylene chloride

OLED device performance

ITO/NPD/Alq3:DMQA (1.8 wt %)/LiF/Al

  • Color: green
  • Max. Luminance: 22000 Cd/m2
  • Max. EQE: 2.1 %

ITO/NPD/Alq3:DMQA (5 wt %)/LiF/Al
  • Color: green
  • Max. Luminance: 13000 Cd/m2
  • Max. EQE: 1.5 %

Application

DMQA may be used to explore the performance of indium tin oxide (ITO)/N,N′-bis-(1-naphthyl)-N,N′-diphenyl-1,1′-biphenyl-4,4′-diamine (NPB)/tris-(8-hydroxyquinolate)-aluminum (Alq3)/MgAg based organic light emitting diodes.[1][2] When used as the emissive dopant in an Alq host layer, DMQA provides improved operational stability.[3]
Green dopant for OLED applications.

General description

Fluorescent dye

Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Doped organic electroluminescent devices with improved stability.
Shi J and Tang CW
Applied Physics Letters null
Electroluminescence of organic light-emitting diodes with an ultra-thin layer of dopant
Li W, et al.
Materials Science and Engineering, B, 77-81 null
Study on electroluminescence processes in dye-doped organic light-emitting diodes
Li W, et al.
Journal of Luminescence null
Study on electroluminescence processes in dye-doped organic light-emitting diodes
Li W, et al.
Journal of Luminescence, 1175-1179 null
Shi, J.; Tang, C.W.
Applied Physics Letters, 70, 1665-1665 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service