558516

Sigma-Aldrich

4,5-Dimethyl-2-isobutyl-3-thiazoline

97% (mixture of isomers)

Empirical Formula (Hill Notation):
C9H17NS
CAS Number:
Molecular Weight:
171.30
EC Number:
MDL number:
PubChem Substance ID:

Quality Level

assay

97% (mixture of isomers)

refractive index

n20/D 1.4870 (lit.)

bp

208 °C/760 mmHg (lit.)

density

1.487 g/mL at 25 °C (lit.)

SMILES string

CC(C)CC1SC(C)C(C)=N1

InChI

1S/C9H17NS/c1-6(2)5-9-10-7(3)8(4)11-9/h6,8-9H,5H2,1-4H3

InChI key

FDOISHJOXPONIV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4,5-Dimethyl-2-isobutyl-3-thiazoline is a flavoring agent.

Packaging

25 g in glass bottle

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

WGK Germany

2

Flash Point(F)

188.6 °F

Flash Point(C)

87 °C

Burdock GA.
Encyclopedia of Food and Color Additives, 860-860 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.