Documents

560561

Sigma-Aldrich

PSS-Octa(2-dichloromethylsilylethyl) substituted

Synonym(s):
Octa(dichlorosilylethyl)-POSS, 1,3,5,7,9,11,13,15-Octa[2-(dichloromethylsilyl)ethyl]pentacyclo-[9.5.1.13,9.15,15.17,13]octasiloxane, POSS-Octa(dichlorosilylethyl) substituted
Empirical Formula (Hill Notation):
C24H56Cl16O12Si16
CAS Number:
Molecular Weight:
1553.31
MDL number:

mp

>300 °C (lit.)

solubility

THF: soluble
acetonitrile: insoluble
chloroform: soluble

SMILES string

C[Si](Cl)(Cl)CC[Si]12O[Si]3(CC[Si](C)(Cl)Cl)O[Si]4(CC[Si](C)(Cl)Cl)O[Si](CC[Si](C)(Cl)Cl)(O1)O[Si]5(CC[Si](C)(Cl)Cl)O[Si](CC[Si](C)(Cl)Cl)(O2)O[Si](CC[Si](C)(Cl)Cl)(O3)O[Si](CC[Si](C)(Cl)Cl)(O4)O5

Legal Information

POSS is a registered trademark of Hybrid Plastics, Inc.

WGK Germany

3

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.