Skip to Content
MilliporeSigma

561185

1-(4-Methylphenyl)-3-(1-pyrrolidinyl)-1-propanone hydrochloride

97%

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C14H19NO · HCl
CAS Number:
Molecular Weight:
253.77
UNSPSC Code:
12352100
PubChem Substance ID:
EC Number:
307-385-7
MDL number:
Assay:
97%
Form:
solid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

97%

form

solid

mp

152-157 °C (lit.)

SMILES string

Cl[H].Cc1ccc(cc1)C(=O)CCN2CCCC2

InChI

1S/C14H19NO.ClH/c1-12-4-6-13(7-5-12)14(16)8-11-15-9-2-3-10-15;/h4-7H,2-3,8-11H2,1H3;1H

InChI key

LZTSEHXSSHJUIV-UHFFFAOYSA-N



Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content