563803

Sigma-Aldrich

5-Chloro-2-thienylmagnesium bromide

0.5 M slurry in THF

Empirical Formula (Hill Notation):
C4H2BrClMgS
CAS Number:
Molecular Weight:
221.79
MDL number:
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

reaction suitability

reaction type: Grignard Reaction

concentration

0.5 M slurry in THF

bp

65 °C (lit.)

density

0.969 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

Clc1ccc(MgBr)s1

InChI

1S/C4H2ClS.BrH.Mg/c5-4-2-1-3-6-4;;/h1-2H;1H;/q;;+1/p-1

InChI key

BTGXLAKETDWIAB-UHFFFAOYSA-M

Related Categories

Packaging

50 mL in Sure/Seal™

Legal Information

Product of Rieke Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.

Signal Word

Danger

Target Organs

Respiratory system

Supp Hazards

EUH014 - EUH019

Hazard Codes

F,C

Risk Statement

11-14-19-34-37-40

Safety Statement

16-26-36/37/39-45

RIDADR

UN 2924 8(3) / PGII

WGK Germany

WGK 3

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.