Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

564605

Sigma-Aldrich

3,3′,5-Triiodo-L-thyronine

95%

Synonym(s):

O-(4-Hydroxy-3-iodophenyl)-3,5-diiodo-L-tyrosine, Liothyronine, T3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12I3NO4
CAS Number:
Molecular Weight:
650.97
Beilstein/REAXYS Number:
2710227
EC Number:
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:

assay

95%

optical activity

[α]20/D +23.0°, c = 2 in ethanol or 0.1 N HCl

mp

234-238 °C (lit.)

SMILES string

N[C@@H](Cc1cc(I)c(Oc2ccc(O)c(I)c2)c(I)c1)C(O)=O

InChI

1S/C15H12I3NO4/c16-9-6-8(1-2-13(9)20)23-14-10(17)3-7(4-11(14)18)5-12(19)15(21)22/h1-4,6,12,20H,5,19H2,(H,21,22)/t12-/m0/s1

InChI key

AUYYCJSJGJYCDS-LBPRGKRZSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

STOT RE 1

target_organs

Thyroid,Cardio-vascular system,Kidney

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Calvo et al.
The Journal of clinical investigation, 86(3), 889-899 (1990-09-01)
To study the protective effects of maternal thyroxine (T4) and 3,5,3'-triiodothyronine (T3) in congenital hypothyroidism, we gave pregnant rats methimazole (MMI), an antithyroid drug that crosses the placenta, and infused them with three different doses of T4 or T3. The
H Mizuma et al.
Circulation research, 88(3), 313-318 (2001-02-17)
Thyroid hormone has been reported to have significant effects on the peripheral vascular system, including relaxation of vascular smooth muscle cells and antiatherosclerotic effects. To exert its biological activity, thyroxine, which is a major secretory product of thyroid gland, needs
E Gunilla Stalenheim
Psychiatry research, 121(3), 281-291 (2003-12-17)
This study is a follow-up investigation of a forensic psychiatric sub-population 6-8 years after forensic psychiatric evaluation. The aim was to examine the long-term validity of biological markers of psychopathy and antisocial behavior over time. Data on criminal records were
Mark D Erion et al.
Proceedings of the National Academy of Sciences of the United States of America, 104(39), 15490-15495 (2007-09-20)
Despite efforts spanning four decades, the therapeutic potential of thyroid hormone receptor (TR) agonists as lipid-lowering and anti-obesity agents remains largely unexplored in humans because of dose-limiting cardiac effects and effects on the thyroid hormone axis (THA), muscle metabolism, and
T Zimmermann-Belsing et al.
Clinical endocrinology, 49(5), 589-595 (1999-04-10)
Previous studies of leptin in thyrotoxic human subjects have been short-term and cross-sectional. We have measured serum leptin concentrations in thyrotoxic patients in order to study the influence of endogenous thyroid hormones on the relationship between serum leptin and fat

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service