571768

Sigma-Aldrich

3,3-Diphenylpropyl isocyanate

97%

Linear Formula:
(C6H5)2CHCH2CH2NCO
CAS Number:
Molecular Weight:
237.30
MDL number:
PubChem Substance ID:
Pricing and availability is not currently available.

assay

97%

mp

51-55 °C (lit.)

SMILES string

O=C=NCCC(c1ccccc1)c2ccccc2

InChI

1S/C16H15NO/c18-13-17-12-11-16(14-7-3-1-4-8-14)15-9-5-2-6-10-15/h1-10,16H,11-12H2

InChI key

BYTUPWXAZYPWKE-UHFFFAOYSA-N

Related Categories

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

UN 3335

WGK Germany

3

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.