Skip to Content
MilliporeSigma

574228

Europium(III) oxalate hydrate

99.999%

Synonym(s):

Ethanedioic acid europium salt hydrate

Sign In to View Organizational & Contract Pricing.

About This Item

Linear Formula:
Eu2(C2O4)3 · xH2O
CAS Number:
Molecular Weight:
567.99 (anhydrous basis)
UNSPSC Code:
12352300
PubChem Substance ID:
EC Number:
221-885-1
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Europium(III) oxalate hydrate, 99.999%

InChI key

DFUMNFRIKCFDGT-UHFFFAOYSA-H

InChI

1S/3C2H2O4.2Eu.H2O/c3*3-1(4)2(5)6;;;/h3*(H,3,4)(H,5,6);;;1H2/q;;;2*+3;/p-6

SMILES string

O.O=C(O[Eu]1OC(=O)C(=O)O1)C(=O)O[Eu]2OC(=O)C(=O)O2

assay

99.999%

reaction suitability

core: europium
reagent type: catalyst

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service