575267

Sigma-Aldrich

Praseodymium(III) bromide hydrate

≥99.99% trace metals basis

Linear Formula:
PrBr3 · xH2O
CAS Number:
Molecular Weight:
380.62 (anhydrous basis)
EC Number:
MDL number:
PubChem Substance ID:
Pricing and availability is not currently available.

assay

≥99.99% trace metals basis

form

chunks

reaction suitability

reagent type: catalyst
core: praseodymium

SMILES string

[H]O[H].Br[Pr](Br)Br

InChI

1S/3BrH.H2O.Pr/h3*1H;1H2;/q;;;;+3/p-3

InChI key

CCMZJPMVOCFMPN-UHFFFAOYSA-K

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

3

Certificate of Analysis
Certificate of Origin
Articles
The rare earth elements impact nearly everyone in the world. All of the people living in advanced technological countries and almost all those living in third world countries utilize the rare earths in their everyday living—the car that one drives (gasoline is refined from oil using rare earth catalysts and catalytic converters reduce the polluting emissions from the automotive exhaust), watching the news on TV (the red and green colors in TV screens), the telephones and computers we use to communicate (the permanent magnets in speakers and disc drives), just to name a few examples.
Read More

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.