579807

Sigma-Aldrich

2,4-Diamino-6-[4-(trifluoromethyl)phenyl]-1,3,5-triazine

98%

Empirical Formula (Hill Notation):
C10H8F3N5
CAS Number:
Molecular Weight:
255.20
MDL number:
PubChem Substance ID:

assay

98%

mp

239-241 °C (lit.)

SMILES string

Nc1nc(N)nc(n1)-c2ccc(cc2)C(F)(F)F

InChI

1S/C10H8F3N5/c11-10(12,13)6-3-1-5(2-4-6)7-16-8(14)18-9(15)17-7/h1-4H,(H4,14,15,16,17,18)

InChI key

BJDQOYKKFRUFII-UHFFFAOYSA-N

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service