Skip to Content
MilliporeSigma

586609

Acetic anhydride-2,2′-13C2,d6

99 atom % 13C, 99 atom % D

Sign In to View Organizational & Contract Pricing.

About This Item

Linear Formula:
(13CD3CO)2O
CAS Number:
Molecular Weight:
110.11
UNSPSC Code:
12352108
PubChem Substance ID:
MDL number:
Isotopic purity:
99 atom % 13C, 99 atom % D
Mass shift:
M+8


isotopic purity

99 atom % 13C, 99 atom % D

bp

138-140 °C (lit.)

mp

-73 °C (lit.)

density

1.164 g/mL at 25 °F

mass shift

M+8

SMILES string

[2H][13C]([2H])([2H])C(=O)OC(=O)[13C]([2H])([2H])[2H]

InChI

1S/C4H6O3/c1-3(5)7-4(2)6/h1-2H3/i1+1D3,2+1D3

InChI key

WFDIJRYMOXRFFG-KLFIIISESA-N

General description

Acetic anhydride-2,2′-13C2,d6 99 atom % 13C, 99 atom % D is a high-purity isotope product with a unique chemical profile that belongs to the class of isotopes. Produced by Sigma-Aldrich, we are committed to providing quality isotope products and a reliable supply chain. Available in industrial and pre-pack quantities.

Application

- High-resolution NMR spectroscopy
- Metabolomic profiling and pathway analysis
- Stable Isotope probing in Environmental microbiology
- Chemical synthesis of labeled compounds

Features and Benefits

Features
  • Versatile use in different formulations involving Acetic anhydride-2,2′-13C2,d6 99 atom % 13C, 99 atom % D.
  • Cost-effective solution for Acetic anhydride-2,2′-13C2,d6 99 atom % 13C, 99 atom % D applications.
  • Exceptional stability and compatibility with various ingredients.
  • High purity of Acetic anhydride-2,2′-13C2,d6 99 atom % 13C, 99 atom % D ensures optimal performance.


Benefits
  • Increases market competitiveness of products containing Acetic anhydride-2,2′-13C2,d6 99 atom % 13C, 99 atom % D.
  • Reduces time to market for products utilizing Acetic anhydride-2,2′-13C2,d6 99 atom % 13C, 99 atom % D.
  • Enhances product efficacy and reliability in formulations.
  • Improves overall product quality and consumer satisfaction.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.


signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

120.2 °F - closed cup

flash_point_c

49 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library