587168

Sigma-Aldrich

Lithium tetrachloroaurate(III)

99.995%

Empirical Formula (Hill Notation):
AuCl4Li
CAS Number:
Molecular Weight:
345.72
PubChem Substance ID:

assay

99.995%

form

solid

SMILES string

[Li+].Cl[Au-](Cl)(Cl)Cl

InChI

1S/Au.4ClH.Li/h;4*1H;/q+3;;;;;+1/p-4

InChI key

AAYAYBKNBYTUKW-UHFFFAOYSA-J

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

3

Certificate of Analysis
Certificate of Origin
Sung Yong Jung et al.
The international journal of cardiovascular imaging, 28(8), 1853-1858 (2012-02-23)
To measure instantaneous velocity fields of venous blood flow in a rat using X-ray particle tracking method. Gold nanoparticles (AuNPs) incorporated chitosan microparticles were applied as biocompatible flow tracers. After intravenous injection of the AuNP-chitosan particles into 7- to 9-week-old...
Limin Yang et al.
Talanta, 89, 136-141 (2012-01-31)
A simple and effective strategy was proposed for synthesis of AuNPs@polyaniline (AuNPs@PANI) core-shell nanocomposites. AuNPs@PANI nanocomposites were prepared by one-step chemical oxidative polymerization of aniline using chloroaurate acid as the oxidant and AuNPs as the seeds. The synthesized AuNPs@PANI nanocomposites...
Sudip Mukherjee et al.
Nanotechnology, 23(45), 455103-455103 (2012-10-16)
The biological approach to synthesis of AuNPs is eco-friendly and an ideal method to develop environmentally sustainable nanoparticles alternative to existing methods. We have developed a simple, fast, clean, efficient, low-cost and eco-friendly single-step green chemistry approach for the synthesis...
Sougata Ghosh et al.
Journal of nanobiotechnology, 10, 17-17 (2012-05-03)
Novel approaches for synthesis of gold nanoparticles (AuNPs) are of utmost importance owing to its immense applications in diverse fields including catalysis, optics, medical diagnostics and therapeutics. We report on synthesis of AuNPs using Gnidia glauca flower extract (GGFE), its...
Qingshuo Chen et al.
Talanta, 89, 142-148 (2012-01-31)
In this work, we utilize the triangular gold nanoparticles (AuNPs) prepared by trisodium citrate reduction of HAuCl(4) in presence of nonionic fluorosurfactant (FSN) as a novel chemiluminescence (CL) probe for the determination of captopril. Captopril can induce a sharp decrease...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.