592803

Sigma-Aldrich

N-(4-Tolyl)formamide

97%

Linear Formula:
HCONHC6H4CH3
CAS Number:
Molecular Weight:
135.16
EC Number:
MDL number:
PubChem Substance ID:

assay

97%

mp

51-55 °C (lit.)

SMILES string

[H]C(=O)Nc1ccc(C)cc1

InChI

1S/C8H9NO/c1-7-2-4-8(5-3-7)9-6-10/h2-6H,1H3,(H,9,10)

InChI key

GRVKDWHXLFEVBP-UHFFFAOYSA-N

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

3

Flash Point(F)

>230 °F

Flash Point(C)

>110 °C

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.