597295

Sigma-Aldrich

3-(Ethylthio)phenylboronic acid

Linear Formula:
H3CCH2SC6H4B(OH)2
CAS Number:
Molecular Weight:
182.05
MDL number:
PubChem Substance ID:

mp

118-122 °C (lit.)

SMILES string

CCSc1cccc(c1)B(O)O

InChI

1S/C8H11BO2S/c1-2-12-8-5-3-4-7(6-8)9(10)11/h3-6,10-11H,2H2,1H3

InChI key

CZODYJDCHKOBID-UHFFFAOYSA-N

Packaging

1 g in glass bottle

Other Notes

Contains varying amounts of anhydride

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service