All Photos(1)

597341

Sigma-Aldrich

2,4-Diamino-6-(2-fluorophenyl)-1,3,5-triazine

97%

Empirical Formula (Hill Notation):
C9H8FN5
CAS Number:
Molecular Weight:
205.19
MDL number:
PubChem Substance ID:

assay

97%

mp

243-247 °C (lit.)

SMILES string

Nc1nc(N)nc(n1)-c2ccccc2F

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service