598062

Sigma-Aldrich

3,5-Dimethyl-4-methoxyphenylboronic acid

≥95%

Synonym(s):
4-Methoxy-3,5-dimethylbenzeneboronic acid, 4-Methoxy-3,5-dimethylphenylboronic acid
Linear Formula:
(CH3)2C6H2OCH3B(OH)2
CAS Number:
Molecular Weight:
180.01
MDL number:
PubChem Substance ID:

assay

≥95%

mp

235-242 °C (lit.)

SMILES string

COc1c(C)cc(cc1C)B(O)O

InChI

1S/C9H13BO3/c1-6-4-8(10(11)12)5-7(2)9(6)13-3/h4-5,11-12H,1-3H3

InChI key

WZUCSPWZHRVOSD-UHFFFAOYSA-N

Application

Reactant for:
  • Preparation of biologically and pharmacologically active molecules
  • Suzuki and Stille cross-coupling reaction
  • Mizoroki-Heck arylation
  • Baeyer-Villiger oxidation
  • Rhodium-catalyzed asymmetric conjugate addition reactions

Packaging

1, 10 g in glass bottle

Other Notes

contains varying amounts of anhydride

RIDADR

NONH for all modes of transport

WGK Germany

3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.