608939

Sigma-Aldrich

Nitromethane-15N

98 atom % 15N, 98% (CP)

Linear Formula:
CH315NO2
Molecular Weight:
62.03
MDL number:
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % 15N

Quality Level

assay

98% (CP)

bp

101 °C (lit.)

density

1.145 g/mL at 25 °C

mass shift

M+1

SMILES string

C[15N+]([O-])=O

InChI

1S/CH3NO2/c1-2(3)4/h1H3/i2+1

InChI key

LYGJENNIWJXYER-VQEHIDDOSA-N

Related Categories

Packaging

1 g in glass bottle
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

hazcat

Acute Tox. 4 Oral - Flam. Liq. 3

storage_class_code

3 - Flammable liquids

WGK Germany

WGK 3

Flash Point(F)

96.8 °F - closed cup

Flash Point(C)

36 °C - closed cup

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service