Skip to Content
MilliporeSigma

Skip To

616591

4-Methoxybenz-3-d1-aldehyde

97 atom % D

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C8DH7O2
Molecular Weight:
137.15
UNSPSC Code:
12352114
PubChem Substance ID:
MDL number:
Isotopic purity:
97 atom % D
Mass shift:
M+1
Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service


isotopic purity

97 atom % D

bp

248 °C (lit.)

density

1.127 g/mL at 25 °C

mass shift

M+1

SMILES string

[2H]c1cc(C=O)ccc1OC

InChI

1S/C8H8O2/c1-10-8-4-2-7(6-9)3-5-8/h2-6H,1H3/i4D

InChI key

ZRSNZINYAWTAHE-QYKNYGDISA-N

General description

4-Methoxybenz-3-d1-aldehyde 97 atom % D is a high-purity isotope product with a unique chemical profile that belongs to the class of isotopes. Produced by Sigma-Aldrich, we are committed to providing quality isotope products and a reliable supply chain. Available in industrial and pre-pack quantities.

Application

- Quality control reference standards
- Pharmaceutical formulation development
- reaction mechanism investigations
- Analytical method development

Features and Benefits

Features
  • High purity of 4-Methoxybenz-3-d1-aldehyde 97 atom % D ensures optimal performance.
  • Cost-effective solution for 4-Methoxybenz-3-d1-aldehyde 97 atom % D applications.
  • Versatile use in different formulations involving 4-Methoxybenz-3-d1-aldehyde 97 atom % D.
  • Exceptional stability and compatibility with various ingredients.


Benefits
  • Improves overall product quality and consumer satisfaction.
  • Reduces time to market for products utilizing 4-Methoxybenz-3-d1-aldehyde 97 atom % D.
  • Enhances product efficacy and reliability in formulations.
  • Increases market competitiveness of products containing 4-Methoxybenz-3-d1-aldehyde 97 atom % D.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
616605608505589861
mass shift

M+1

mass shift

M+1

mass shift

M+2

mass shift

M+1

isotopic purity

97 atom % D

isotopic purity

98 atom % D

isotopic purity

99 atom % 13C, 98 atom % D

isotopic purity

98 atom % D

density

1.127 g/mL at 25 °C

density

1.127 g/mL at 25 °C

density

1.135 g/mL at 25 °C

density

1.127 g/mL at 25 °C

bp

248 °C (lit.)

bp

248 °C (lit.)

bp

248 °C (lit.)

bp

248 °C (lit.)




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters