635073

Sigma-Aldrich

Magnesium aluminate

single crystal substrate, <100>, 99.99% trace metals basis

Linear Formula:
MgO·Al2O3
CAS Number:
Molecular Weight:
142.27
EC Number:
MDL number:
PubChem Substance ID:

Quality Level

description

single side polished

assay

99.99% trace metals basis

hardness

8.0 mohs

L × W × thickness

10 mm × 10 mm × 0.5 mm

mp

2130 °C (lit.)

density

3.64 g/mL at 25 °C (lit.)

semiconductor properties

<100>

SMILES string

[Mg++].[O-][Al]=O.[O-][Al]=O

InChI

1S/2Al.Mg.4O/q;;+2;;;2*-1

InChI key

VPBIQXABTCDMAU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Transparent substrate used for bulk microwave and acoustic wave devices. Good substrate for III-V nitride materials. Inert matrix for the transmutation of minor actinides such as americium.

Packaging

1 ea in rigid mailer

Physical properties

propagation loss: 6.5 dB/μs; phase velocity: 6500 m/x at (100> shear wave); thermal expansion: 7.45 × 10-6 /°C

Physical form

cubic (a = 8.083 Å)

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

3

Certificate of Analysis
Certificate of Origin
Klaassen, F.C.
J. Nuc. Mat., 319, 108-108 (2003)
Satit Puttipipatkhachorn et al.
International journal of pharmaceutics, 293(1-2), 51-62 (2005-03-22)
Diclofenac calcium-alginate (DCA) beads were reinforced with different amounts of sodium starch glycolate (SSG) or magnesium aluminum silicate (MAS) and were prepared using ionotropic gelation method. Complex formation of sodium alginate (SA) and SSG or MAS in calcium-alginate beads was...
J C Keller et al.
Dental materials : official publication of the Academy of Dental Materials, 13(1), 62-68 (1997-01-01)
The objective of this work was to examine osteoblast-like cell attachment and morphology in vitro to osteoceramic materials with three different surface morphologies. Osteoceramic composite disks were fabricated from tricalcium phosphate and magnesium-aluminate spinel (MgAl2O4) in a 50 vol% ratio....
Tomohito Kameda et al.
Chemosphere, 73(5), 844-847 (2008-07-25)
Magnesium-aluminum oxide (Mg-Al oxide) obtained by thermal decomposition of Mg-Al layered double hydroxide (Mg-Al LDH) effectively removed HCl from gaseous streams. HCl removal was greater in the presence of added water vapor at all temperatures examined and increased with decreasing...
Gabriel A Devenyi et al.
Nano letters, 9(12), 4258-4263 (2009-10-22)
A new class of gold nanostructures has been fabricated on the (100), (111), and (110) surfaces of lattice-matched MgAl(2)O(4) substrates. The nanostructures were fabricated through a synthesis route where a thin gold film dewets, liquefies, and then slowly self-assembles. The...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.