Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|---|---|---|
| 1 g | Available to ship TODAYfromMILWAUKEE | $840.00 $630.00 |
About This Item
Linear Formula:
[NH2(CH2)4NH2]:(G=5);dendri PAMAM(NHCOCH2CH2COOH)128
Molecular Weight:
41662.18
UNSPSC Code:
12162002
NACRES:
NA.23
MDL number:
$630.00
List Price$840.00Save 25%Web-Only Promotion
Available to ship TODAYDetails
description
succinamic acid surface groups
Quality Level
form
solution
mol wt
generation 5
feature
core type 1,4-diaminobutane core (4-carbon core)
concentration
10 wt. % in H2O
no. Surface Groups
128
storage temp.
−20°C
Features and Benefits
Contains 128 surface succinamic acid groups.
Unlike the cationic amine surface functionalized dendrimers, succinamic acid functional dendrimers offer the researcher anionic surface groups, typical carboxylic acid reactivity and the potential to make carboxylate salts for a variety of reaction capabilities.
Packaging
unit weight includes solvent
Legal Information
Mfg. by Dendritic NanoTechnologies Inc. Licensed for research use only.
1 of 1
This Item | |||
|---|---|---|---|
| concentration 10 wt. % in H2O | concentration 10 wt. % in H2O | concentration 5 wt. % in methanol | concentration 20 wt. % in methanol |
| storage temp. −20°C | storage temp. −20°C | storage temp. - | storage temp. 2-8°C |
| form solution | form solution | form liquid | form liquid |
| mol wt generation 5 | mol wt generation 4 | mol wt generation 4.5 | mol wt generation 0.5 |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| description succinamic acid surface groups | description succinamic acid surface groups | description sodium carboxylate surface groups | description sodium carboxylate surface groups |
Storage Class
10 - Combustible liquids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 635898-1G | 04061826751435 |



