MilliporeSigma
All Photos(1)

643971

Sigma-Aldrich

3-(3-Hydroxyphenyl)benzonitrile

97%

Synonym(s):
3′-Hydroxy-[1,1′−biphenyl]-3-carbonitrile, 3′-Hydroxybiphenyl-3-carbonitrile
Empirical Formula (Hill Notation):
C13H9NO
CAS Number:
Molecular Weight:
195.22
MDL number:
PubChem Substance ID:
NACRES:
NA.22

Quality Level

assay

97%

mp

105-109 °C (lit.)

SMILES string

Oc1cccc(c1)-c2cccc(c2)C#N

InChI

1S/C13H9NO/c14-9-10-3-1-4-11(7-10)12-5-2-6-13(15)8-12/h1-8,15H

InChI key

IYDCWWPOHIRIQG-UHFFFAOYSA-N

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service