All Photos(1)

645842

Sigma-Aldrich

3-Bromo-2-(4′-chlorobenzyloxy)phenylboronic acid

Empirical Formula (Hill Notation):
C13H11BBrClO3
CAS Number:
Molecular Weight:
341.39
MDL number:
PubChem Substance ID:

mp

104-109 °C (lit.)

SMILES string

OB(O)c1cccc(Br)c1OCc2ccc(Cl)cc2

InChI

1S/C13H11BBrClO3/c15-12-3-1-2-11(14(17)18)13(12)19-8-9-4-6-10(16)7-5-9/h1-7,17-18H,8H2

InChI key

KBWXXELCTOFTRW-UHFFFAOYSA-N

Packaging

1 g in glass bottle

Other Notes

Contains varying amounts of anhydride

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service