Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C5H6N2OS
CAS Number:
Molecular Weight:
142.18
UNSPSC Code:
12352100
PubChem Substance ID:
EC Number:
220-321-1
MDL number:
assay
98%
form
solid
mp
202-206 °C (dec.) (lit.)
SMILES string
CC(=O)Nc1nccs1
InChI
1S/C5H6N2OS/c1-4(8)7-5-6-2-3-9-5/h2-3H,1H3,(H,6,7,8)
InChI key
WXPLRSVMGRAIGW-UHFFFAOYSA-N
Storage Class
13 - Non Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.