650862

Sigma-Aldrich

(R)-C3-TunePhos

Synonym(s):
(R)-1,13-Bis(diphenylphosphino)-7,8-dihydro-6H-dibenzo[f,h][1,5]dioxonin
Empirical Formula (Hill Notation):
C39H32O2P2
CAS Number:
Molecular Weight:
594.62
MDL number:
PubChem Substance ID:

optical activity

[α]20/D -252±5°, c = 1 in chloroform

mp

153-161 °C (lit.)

SMILES string

C1COc2cccc(P(c3ccccc3)c4ccccc4)c2-c5c(OC1)cccc5P(c6ccccc6)c7ccccc7

InChI

1S/C39H32O2P2/c1-5-16-30(17-6-1)42(31-18-7-2-8-19-31)36-26-13-24-34-38(36)39-35(41-29-15-28-40-34)25-14-27-37(39)43(32-20-9-3-10-21-32)33-22-11-4-12-23-33/h1-14,16-27H,15,28-29H2

InChI key

GTIXSUJKFAATAE-UHFFFAOYSA-N

Application

Chiral Quest Phosphine Ligands for Asymmetric Hydrogenation

Ligand used in:
  • Redox-neutral domino reaction of arylethynylbenzylidenearylpentynones catalyzed by gold catalyst
  • Asymmetric allylic O-alkylation
  • Asymmetric alkylation of racemic secondary phosphines catalyzed by ruthenium chiral diphosphine hydride complexes
  • Stereoselective preparation of cyclopropylcarboxaldehydes via Rh-catalyzed asymmetric hydroformylation of cyclopropenes
  • Rhodium-catalyzed asymmetric hydrogenations
Provides comparable or superior enantioselectivities and catalytic abilities to BINAP in Ru-catalyzed asymmetric hydrogenations of β-keto esters, cyclic β-(acylamino)acrylates and α-phthalimide ketones.

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Certificate of Origin

Aiwen Lei et al.
Journal of the American Chemical Society, 126(6), 1626-1627 (2004-02-12)
A new type of alpha-phthalimide ketones was hydrogenated in excellent enantioselectivity by using a Ru-(C3-TunePhos) complex as the catalyst. Up to 10 000 turnovers have been achieved in more than 99% ee in the hydrogenation reaction. A dynamic kinetic resolution...
Wenjun Tang et al.
Journal of the American Chemical Society, 125(32), 9570-9571 (2003-08-09)
Hydrogenation of a series of cyclic beta-(acylamino)acrylates with tetrasubstituted olefins has been accomplished successfully with the use of Ru catalysts with chiral biaryl ligands such as C3-TunaPhos, and up to over 99% ee's have been achieved. This methodology provides an...
Synthesis of chiral bisphosphines with tunable bite angles and their applications in asymmetric hydrogenation of beta-ketoesters.
Z Zhang et al.
The Journal of organic chemistry, 65(19), 6223-6226 (2000-09-16)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.