652466

Sigma-Aldrich

Strontium fluoride

precipitated, 99.995% trace metals basis

Synonym(s):
Strontium difluoride
Linear Formula:
SrF2
CAS Number:
Molecular Weight:
125.62
EC Number:
MDL number:
PubChem Substance ID:

Quality Level

assay

99.995% trace metals basis

form

solid

quality

precipitated

impurities

≤ 55.0  ppm Trace Metal Analysis

bp

2489 °C (lit.)

mp

>1400 °C (lit.)

density

4.24 g/mL at 25 °C (lit.)

SMILES string

F[Sr]F

InChI

1S/2FH.Sr/h2*1H;/q;;+2/p-2

InChI key

FVRNDBHWWSPNOM-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Certificate of Origin

Kan Usuda et al.
Biological trace element research, 167(1), 84-90 (2015-03-10)
Various fluoride compounds are widely used in industry. The present risk assessment study was conducted using a series of inorganic binary fluorides of the type XFn, where X(n) = Na(+), K(+), Li(+), Mg(2+), Ca(2+), Sr(2+), Ba(2+), Al(3+), Nd(3+), La(3+), Ce(3+), Sm(3+), Gd(3+)...
Bikramjit Basu et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 103(6), 1168-1179 (2014-10-11)
One of the desired properties for any new biomaterial composition is its long-term stability in a suitable animal model and such property cannot be appropriately assessed by performing short-term implantation studies. While hydroxyapatite (HA) or bioglass coated metallic biomaterials are...
Articles
Spectral conversion for solar cells is an emerging concept in the field of photovoltaics, and it has the potential to increase significantly the efficiency of solar cells. Lanthanide ions are ideal candidates for spectral conversion, due to their high luminescence efficiencies and rich energy level structure that allows for great flexibility in the upconversion and downconversion of photons in a wide spectral region (NIR-VIS-UV).
Read More

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.