653454

Sigma-Aldrich

5-Bromo-2-benzoxazolinone

97%

Synonym(s):
5-Bromo-2(3H)-benzoxazolone
Empirical Formula (Hill Notation):
C7H4BrNO2
CAS Number:
Molecular Weight:
214.02
MDL number:
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

assay

97%

mp

218-222 °C (lit.)

SMILES string

Brc1ccc2OC(=O)Nc2c1

InChI

1S/C7H4BrNO2/c8-4-1-2-6-5(3-4)9-7(10)11-6/h1-3H,(H,9,10)

InChI key

DMHTZWJRUUOALC-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

Hazard Codes

Xn

Risk Statement

22-36

Safety Statement

26

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.