All Photos(1)

669563

Sigma-Aldrich

Boc-Phe(3,5-F2)-OH

≥98.0% (HPLC)

Synonym(s):
Boc-3,5-difluoro-L-phenylalanine
Empirical Formula (Hill Notation):
C14H17F2NO4
CAS Number:
Molecular Weight:
301.29
MDL number:
PubChem Substance ID:
NACRES:
NA.22

Quality Level

assay

≥98.0% (HPLC)

reaction suitability

reaction type: Boc solid-phase peptide synthesis

application(s)

peptide synthesis: suitable

SMILES string

CC(C)(C)OC(=O)N[C@@H](Cc1cc(F)cc(F)c1)C(O)=O

InChI

1S/C14H17F2NO4/c1-14(2,3)21-13(20)17-11(12(18)19)6-8-4-9(15)7-10(16)5-8/h4-5,7,11H,6H2,1-3H3,(H,17,20)(H,18,19)/t11-/m0/s1

InChI key

CZBNUDVCRKSYDG-NSHDSACASA-N

Storage Class Code

13 - Non Combustible Solids

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

Certificate of Analysis

Certificate of Origin

Articles

Unnatural Amino Acids

Unnatural amino acids, the non-proteinogenic amino acids that either occur naturally or are chemically synthesized, are becoming more and more important as tools for modern drug discovery research.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service