Skip to Content
MilliporeSigma

678198

Cefaclor-(phenyl-13C6)

99 atom % 13C

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
13C6C9H14ClN3O4S·H2O
Molecular Weight:
391.78
PubChem Substance ID:
UNSPSC Code:
12352005
Isotopic purity:
99 atom % 13C
Mass shift:
M+6
Form:
solid


isotopic purity

99 atom % 13C

form

solid

mol wt

391.72 by atom % calculation

technique(s)

mass spectrometry (MS): suitable

mass shift

M+6

SMILES string

ClC1=C(C(O)=O)N2[C@@H]([C@H](NC([C@H]([13C]3=[13CH][13CH]=[13CH][13CH]=[13CH]3)N)=O)C2=O)SC1

InChI

1S/C15H14ClN3O4S/c16-8-6-24-14-10(13(21)19(14)11(8)15(22)23)18-12(20)9(17)7-4-2-1-3-5-7/h1-5,9-10,14H,6,17H2,(H,18,20)(H,22,23)/t9-,10+,14+/m0/s1/i1+1,2+1,3+1,4+1,5+1,7+1

InChI key

QYIYFLOTGYLRGG-VSDCQUGNSA-N

General description

Cefaclor-(phenyl-13C6) 99 atom % 13C is a high-purity isotope product with a unique chemical profile that belongs to the class of isotopes. Produced by Sigma-Aldrich, we are committed to providing quality isotope products and a reliable supply chain. Available in industrial and pre-pack quantities.

Application

- High-resolution NMR spectroscopy
- Metabolomic profiling and pathway analysis
- Stable Isotope probing in Environmental microbiology
- reaction mechanism investigations

Features and Benefits

Features
  • Cost-effective solution for Cefaclor-(phenyl-13C6) 99 atom % 13C applications.
  • High purity of Cefaclor-(phenyl-13C6) 99 atom % 13C ensures optimal performance.
  • Versatile use in different formulations involving Cefaclor-(phenyl-13C6) 99 atom % 13C.
  • Exceptional stability and compatibility with various ingredients.


Benefits
  • Enhances product efficacy and reliability in formulations.
  • Reduces time to market for products utilizing Cefaclor-(phenyl-13C6) 99 atom % 13C.
  • Increases market competitiveness of products containing Cefaclor-(phenyl-13C6) 99 atom % 13C.
  • Improves overall product quality and consumer satisfaction.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library