Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C50H32FeO4P2
CAS Number:
Molecular Weight:
814.58
UNSPSC Code:
12352000
PubChem Substance ID:
MDL number:
SMILES string
[Fe].[CH]1[CH][CH][C]([CH]1)P2Oc3ccc4ccccc4c3-c5c(O2)ccc6ccccc56.[CH]7[CH][CH][C]([CH]7)P8Oc9ccc%10ccccc%10c9-c%11c(O8)ccc%12ccccc%11%12
InChI
1S/2C25H16O2P.Fe/c2*1-5-11-20-17(7-1)13-15-22-24(20)25-21-12-6-2-8-18(21)14-16-23(25)27-28(26-22)19-9-3-4-10-19;/h2*1-16H;
InChI key
WMKZSCONNPWJGW-UHFFFAOYSA-N
Application
Used in a variety of asymmetric reactions, including: hydrogenations and conjugate additions.
Storage Class
13 - Non Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Reetz, M. T. et al.
Chemical Communications (Cambridge, England), 2077-2077 (1998)
Reetz, M. T.
Pure and Applied Chemistry. Chimie Pure Et Appliquee, 71, 1503-1503 (1999)