68602

Sigma-Aldrich

cis-2- Amino-3-cyclopentene-1-carboxylic acid hydrochloride

≥97.0% (AT)

Empirical Formula (Hill Notation):
C6H9NO2 · HCl
CAS Number:
Molecular Weight:
163.60
MDL number:
PubChem Substance ID:

assay

≥97.0% (AT)

application(s)

peptide synthesis: suitable

SMILES string

Cl.NC1C=CCC1C(O)=O

InChI

1S/C6H9NO2.ClH/c7-5-3-1-2-4(5)6(8)9;/h1,3-5H,2,7H2,(H,8,9);1H

InChI key

NJIAFLWOIUSOSY-UHFFFAOYSA-N

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service