688371

Sigma-Aldrich

1,4-Dioxane solution

NMR reference standard, 10 mM in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.

Empirical Formula (Hill Notation):
C4H8O2
CAS Number:
Molecular Weight:
88.11
EC Number:
MDL number:
PubChem Substance ID:
NACRES:
NA.12
Pricing and availability is not currently available.

Quality Level

grade

NMR reference standard

concentration

10 mM in methanol-d4 (99.8 atom % D)

application(s)

NMR: suitable

NMR tube size

5 mm × 8 in.

SMILES string

C1COCCO1

InChI

1S/C4H8O2/c1-2-6-4-3-5-1/h1-4H2

InChI key

RYHBNJHYFVUHQT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Features and Benefits

13C sensitivity

Quantity

5 mm O.D. tube contains 0.700 mL.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Codes

F,T

Risk Statement

11-23/24/25-39/23/24/25

Safety Statement

7-16-36/37-45

RIDADR

UN1230 - class 3 - PG 2 - Methanol, solution

WGK Germany

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.